Assay Details
| Gene Symbol: Mmp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008611.4 | NP_032637.3 | 7 - 8 | 1147 | 94 | |
| GenBank mRNA | AK089234.1 | - | 7 - 8 | 1118 | 94 | |
| AK089336.1 | - | 7 - 8 | 1122 | 94 | ||
| AK089381.1 | - | 7 - 8 | 1119 | 94 | ||
| AK089516.1 | - | 7 - 8 | 1108 | 94 | ||
| AK137468.1 | - | 7 - 8 | 1119 | 94 | ||
| AK154937.1 | - | 7 - 8 | 1118 | 94 | ||
| AK155740.1 | - | 7 - 8 | 1118 | 94 | ||
| AK155901.1 | - | 7 - 8 | 1122 | 94 | ||
| AK170177.1 | - | 7 - 8 | 1118 | 94 | ||
| AK170388.1 | - | 7 - 8 | 1120 | 94 | ||
| AK171168.1 | - | 7 - 8 | 1118 | 94 | ||
| AK171613.1 | - | 7 - 8 | 1120 | 94 | ||
| BC042742.1 | - | 7 - 8 | 1128 | 5032770 | 94 | |
| U96696.1 | - | 7 - 8 | 1113 | 94 | ||
| Y13342.1 | - | 7 - 8 | 1045 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp8
Genomic Map