Assay Details
| Gene Symbol: Kif23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024245.4 | NP_077207.3 | 2 - 3 | 256 | 115 | |
| GenBank mRNA | AK013939.1 | - | 2 - 3 | 194 | 115 | |
| AK030771.1 | - | 2 - 3 | 233 | 115 | ||
| BC003363.1 | - | 2 - 3 | 229 | 115 | ||
| BC047273.1 | - | 2 - 3 | 214 | 6440797 | 115 | |
| BY132148.1 | - | 2 - 3 | 256 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif23
Genomic Map