Assay Details
| Gene Symbol: Tecta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009347.2 | NP_033373.2 | 18 - 19 | 5848 | 95 | |
| GenBank mRNA | AK136719.1 | - | 3 - 4 | 674 | 95 | |
| BC138426.1 | - | 18 - 19 | 5631 | 8861446 | 95 | |
| BC171953.1 | - | 18 - 19 | 5631 | 9054239 | 95 | |
| X99805.1 | - | 18 - 19 | 5848 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tecta
Genomic Map