Assay Details
| Gene Symbol: Tecta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009347.2 | NP_033373.2 | 1 - 2 | 269 | 109 | |
| GenBank mRNA | BC138426.1 | - | 1 - 2 | 67 | 8861446 | 109 |
| BC171953.1 | - | 1 - 2 | 67 | 9054239 | 109 | |
| X99805.1 | - | 1 - 2 | 269 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138426.1, BC171953.1, NM_009347.2, X99805.1 transcripts.
Set Membership: |
|

Tecta
Genomic Map