Assay Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110251.1 | NP_001103721.1 | 10 - 11 | 778 | 81 | |
| NM_013551.2 | NP_038579.2 | 10 - 11 | 836 | 81 | ||
| GenBank mRNA | AK028234.1 | - | 10 - 11 | 704 | 81 | |
| AK143558.1 | - | 10 - 11 | 703 | 81 | ||
| AK151020.1 | - | 10 - 11 | 802 | 81 | ||
| AK166734.1 | - | 10 - 11 | 734 | 81 | ||
| AK167702.1 | - | 10 - 11 | 795 | 81 | ||
| AK218865.1 | - | 1 - 2 | 50 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
heme biosynthetic process
peptidyl-pyrromethane cofactor linkage
tetrapyrrole biosynthetic process
hydroxymethylbilane synthase activity
uroporphyrinogen-III synthase activity
transferase activity
carboxylic acid binding
amine binding
coenzyme binding
porphyrin-containing compound biosynthetic process
heme biosynthetic process
peptidyl-pyrromethane cofactor linkage
tetrapyrrole biosynthetic process
hydroxymethylbilane synthase activity
uroporphyrinogen-III synthase activity
transferase activity
carboxylic acid binding
amine binding
coenzyme binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467130 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467132 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988102 | AK167702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988102 | AK151020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988102 | AK153884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988102 | AK166734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988102 | NM_013551.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31260 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988102 | AK167702.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988102 | AK151020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988102 | AK153884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988102 | AK166734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988102 | NM_013551.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992487 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155361 | AK167702.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155361 | BC003861.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155361 | AK153884.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155361 | AK166734.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155361 | NM_013551.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26467137 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06769182 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01918405 | - | - |
| Whole Mouse Genome G4122A | A_52_P578181 | AK158160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155361 | AK167702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155361 | BC003861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155361 | AK153884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155361 | AK166734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155361 | NM_013551.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01918405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07646520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07604766 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07646523 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467130 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26467132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26467120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07549889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07734133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07646520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07887522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07868334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08050960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08088914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26467116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592823 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426475_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10350335 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592816 | - | - |
| Mouse Genome 430 2.0 Array | 1426475_at | - | - |
| Mouse Genome 430 2.0 Array | 1426475_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436930_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436930_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15288_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15288_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900017921.mm.1 | NM_001110251.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017921.mm.1 | NM_001110251.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017921.mm.1 | NM_013551.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017921.mm.1 | NM_013551.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017921.mm.1 | TC0900002158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900017920.mm.1 | NM_001110251.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017920.mm.1 | NM_001110251.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017920.mm.1 | NM_013551.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017920.mm.1 | NM_013551.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017920.mm.1 | TC0900002158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900017922.mm.1 | NM_001110251.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017922.mm.1 | NM_001110251.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017922.mm.1 | NM_013551.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017922.mm.1 | NM_013551.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017922.mm.1 | TC0900002158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900017923.mm.1 | NM_001110251.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017923.mm.1 | NM_001110251.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017923.mm.1 | NM_013551.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017923.mm.1 | NM_013551.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017923.mm.1 | TC0900002158.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436930_x_at | - | - |
| Mouse Expression Set 430 | 1426475_at | - | - |
| Mouse Expression Set 430 | 1436930_PM_x_at | - | - |
| Mouse Expression Set 430 | 1426475_PM_at | - | - |
| Murine Genome U74v2 Set | 93258_at | - | - |
| Murine Genome U74v2 Set | 93258_PM_at | - | - |
| Murine Genome U74v2 Set | 167513_PM_i_at | - | - |
| Murine Genome U74v2 Set | 167513_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426475_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426475_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436930_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436930_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426475_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426475_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4590605 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1740022 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6650072 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4590605 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1740022 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6650072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769525 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657124 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250541 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2769525 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2657124 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250541 | - | - |
| mouse-6 v1.1 (Illumina) | scl35959.8.1_6-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003429.1_37-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650072 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590605 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35959.8.1_6-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003429.1_37-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740022 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650072 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250541 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657124 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250541 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003429.1_37-S | - | - |

Hmbs
Genomic Map