Assay Details
| Gene Symbol: Phldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153537.4 | NP_705765.3 | 3 - 4 | 277 | 66 | |
| GenBank mRNA | BC058712.1 | - | 3 - 4 | 273 | 6842596 | 66 |
| BY230078.1 | - | 3 - 4 | 278 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phldb1
Genomic Map