Assay Details
| Gene Symbol: Phldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153537.4 | NP_705765.3 | 7 - 8 | 2216 | 105 | |
| GenBank mRNA | AK122336.1 | - | 4 - 5 | 1934 | 105 | |
| BC053931.1 | - | 2 - 3 | 356 | 105 | ||
| BC058712.1 | - | 7 - 8 | 2212 | 6842596 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phldb1
Genomic Map