Assay Details
| Gene Symbol: Usp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016808.2 | NP_058088.2 | 10 - 11 | 1688 | 70 | |
| NM_198091.2 | NP_932759.1 | 9 - 10 | 1144 | 70 | ||
| NM_198092.2 | NP_932760.2 | 10 - 11 | 1785 | 70 | ||
| GenBank mRNA | AB041799.1 | - | 9 - 10 | 1105 | 70 | |
| AF079565.1 | - | 9 - 10 | 746 | 70 | ||
| AK138799.1 | - | 11 - 12 | 1986 | 70 | ||
| AY255637.1 | - | 9 - 10 | 1113 | 70 | ||
| AY255638.1 | - | 10 - 11 | 1667 | 70 | ||
| AY255639.1 | - | 10 - 11 | 1621 | 70 | ||
| AY255640.1 | - | 10 - 11 | 1666 | 70 | ||
| BC017517.1 | - | 10 - 11 | 1667 | 4506362 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp2
Genomic Map