Assay Details
| Gene Symbol: Mmp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008607.2 | NP_032633.1 | 2 - 3 | 407 | 92 | |
| GenBank mRNA | AK150728.1 | - | 2 - 3 | 394 | 92 | |
| AK151626.1 | - | 2 - 3 | 394 | 92 | ||
| AK152507.1 | - | 2 - 3 | 393 | 92 | ||
| AK152916.1 | - | 2 - 3 | 394 | 92 | ||
| AK153458.1 | - | 2 - 3 | 392 | 92 | ||
| AK155188.1 | - | 2 - 3 | 393 | 92 | ||
| AK159921.1 | - | 2 - 3 | 394 | 92 | ||
| AK160819.1 | - | 2 - 3 | 392 | 92 | ||
| BC125320.1 | - | 2 - 3 | 407 | 40129835 | 92 | |
| BC125322.1 | - | 2 - 3 | 407 | 40129837 | 92 | |
| X66473.1 | - | 2 - 3 | 376 | 92 | ||
More Information
Set Membership: |
|

Mmp13
Genomic Map