Assay Details
| Gene Symbol: Mmp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008605.3 | NP_032631.3 | 3 - 4 | 559 | 115 | |
| GenBank mRNA | AK089295.1 | - | 528 | 115 | ||
| AK089309.1 | - | 3 - 4 | 528 | 115 | ||
| AK089335.1 | - | 3 - 4 | 524 | 115 | ||
| AK089452.1 | - | 3 - 4 | 528 | 115 | ||
| AK089509.1 | - | 4 - 5 | 402 | 115 | ||
| AK089523.1 | - | 3 - 4 | 528 | 115 | ||
| AK090051.1 | - | 3 - 4 | 530 | 115 | ||
| AK150596.1 | - | 3 - 4 | 528 | 115 | ||
| AK151021.1 | - | 3 - 4 | 528 | 115 | ||
| AK151273.1 | - | 3 - 4 | 528 | 115 | ||
| AK151933.1 | - | 3 - 4 | 528 | 115 | ||
| AK155139.1 | - | 3 - 4 | 527 | 115 | ||
| AK155743.1 | - | 3 - 4 | 528 | 115 | ||
| AK155747.1 | - | 3 - 4 | 528 | 115 | ||
| AK155748.1 | - | 3 - 4 | 528 | 115 | ||
| AK155841.1 | - | 3 - 4 | 586 | 115 | ||
| AK155865.1 | - | 3 - 4 | 527 | 115 | ||
| AK170360.1 | - | 3 - 4 | 528 | 115 | ||
| AK170364.1 | - | 3 - 4 | 528 | 115 | ||
| AK170500.1 | - | 4 - 5 | 402 | 115 | ||
| AK170667.1 | - | 2 - 3 | 1575 | 115 | ||
| AK170798.1 | - | 3 - 4 | 527 | 115 | ||
| AK170804.1 | - | 3 - 4 | 527 | 115 | ||
| AK170976.1 | - | 3 - 4 | 528 | 115 | ||
| AK171006.1 | - | 3 - 4 | 528 | 115 | ||
| AK171036.1 | - | 3 - 4 | 528 | 115 | ||
| AK171176.1 | - | 3 - 4 | 528 | 115 | ||
| AK171428.1 | - | 3 - 4 | 528 | 115 | ||
| AK171435.1 | - | 2 - 3 | 1575 | 115 | ||
| AK171447.1 | - | 3 - 4 | 528 | 115 | ||
| AK171448.1 | - | 3 - 4 | 528 | 115 | ||
| AK171449.1 | - | 3 - 4 | 528 | 115 | ||
| AK171450.1 | - | 3 - 4 | 528 | 115 | ||
| AK171466.1 | - | 3 - 4 | 528 | 115 | ||
| AK171468.1 | - | 3 - 4 | 528 | 115 | ||
| AK171470.1 | - | 3 - 4 | 528 | 115 | ||
| AK171472.1 | - | 3 - 4 | 528 | 115 | ||
| AK171485.1 | - | 3 - 4 | 528 | 115 | ||
| AK171518.1 | - | 3 - 4 | 528 | 115 | ||
| AK171542.1 | - | 3 - 4 | 528 | 115 | ||
| AK171570.1 | - | 3 - 4 | 528 | 115 | ||
| BC019135.1 | - | 3 - 4 | 522 | 5037674 | 115 | |
| CT010377.1 | - | 3 - 4 | 475 | 115 | ||
| M82831.1 | - | 3 - 4 | 475 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp12
Genomic Map