Assay Details
| Gene Symbol: Mmp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008605.3 | NP_032631.3 | 7 - 8 | 1109 | 104 | |
| GenBank mRNA | AK089309.1 | - | 7 - 8 | 1078 | 104 | |
| AK089452.1 | - | 7 - 8 | 1078 | 104 | ||
| AK089509.1 | - | 8 - 9 | 952 | 104 | ||
| AK089523.1 | - | 7 - 8 | 1078 | 104 | ||
| AK090051.1 | - | 7 - 8 | 1080 | 104 | ||
| AK150596.1 | - | 7 - 8 | 1078 | 104 | ||
| AK151021.1 | - | 7 - 8 | 1078 | 104 | ||
| AK151273.1 | - | 7 - 8 | 1078 | 104 | ||
| AK151933.1 | - | 7 - 8 | 1078 | 104 | ||
| AK155139.1 | - | 7 - 8 | 1077 | 104 | ||
| AK155743.1 | - | 7 - 8 | 1078 | 104 | ||
| AK155747.1 | - | 7 - 8 | 1078 | 104 | ||
| AK155748.1 | - | 7 - 8 | 1078 | 104 | ||
| AK155841.1 | - | 7 - 8 | 1136 | 104 | ||
| AK155865.1 | - | 7 - 8 | 1077 | 104 | ||
| AK170360.1 | - | 7 - 8 | 1078 | 104 | ||
| AK170364.1 | - | 7 - 8 | 1078 | 104 | ||
| AK170500.1 | - | 8 - 9 | 952 | 104 | ||
| AK170798.1 | - | 7 - 8 | 1077 | 104 | ||
| AK170804.1 | - | 7 - 8 | 1077 | 104 | ||
| AK170976.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171006.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171032.1 | - | 7 - 8 | 1077 | 104 | ||
| AK171036.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171176.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171428.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171435.1 | - | 6 - 7 | 2125 | 104 | ||
| AK171447.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171448.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171449.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171450.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171466.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171468.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171470.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171472.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171485.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171518.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171542.1 | - | 7 - 8 | 1078 | 104 | ||
| AK171570.1 | - | 7 - 8 | 1078 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp12
Genomic Map