Assay Details
| Gene Symbol: Spock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052994.2 | NP_443720.1 | 5 - 6 | 683 | 78 | |
| GenBank mRNA | AJ278999.1 | - | 5 - 6 | 685 | 78 | |
| AK122233.1 | - | 5 - 6 | 345 | 78 | ||
| AK140483.1 | - | 5 - 6 | 589 | 78 | ||
| AK162941.1 | - | 5 - 6 | 588 | 78 | ||
| BC057324.1 | - | 5 - 6 | 683 | 6821449 | 78 | |
| BC060210.1 | - | 5 - 6 | 635 | 6838067 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spock2
Genomic Map