Assay Details
| Gene Symbol: Ccdc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111121.1 | NP_001104591.1 | 5 - 6 | 1007 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc6| Gene Symbol: Ccdc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111121.1 | NP_001104591.1 | 5 - 6 | 1007 | 59 | |
Set Membership: |
|