Assay Details
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081412.2 | NP_001074881.1 | 12 - 13 | 2731 | 76 | |
GenBank mRNA | AK129482.1 | - | 9 - 10 | 921 | 76 | |
AK183908.1 | - | 2 - 3 | 102 | 76 | ||
BC060270.1 | - | 12 - 13 | 1969 | 76 | ||
BC131682.1 | - | 13 - 14 | 2136 | 40111128 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
