Assay Details
| Gene Symbol: Elane | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015779.2 | NP_056594.2 | 1 - 2 | 94 | 69 | |
| GenBank mRNA | AK143710.1 | - | 1 - 2 | 94 | 69 | |
| BC145800.1 | - | 1 - 2 | 159 | 40131103 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elane
Genomic Map