Assay Details
| Gene Symbol: Mum1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023431.5 | NP_075920.4 | 2 - 3 | 223 | 160 | |
| GenBank mRNA | AK154610.1 | - | 2 - 3 | 142 | 160 | |
| AK158750.1 | - | 2 - 3 | 223 | 160 | ||
| AK159972.1 | - | 2 - 3 | 148 | 160 | ||
| AK170459.1 | - | 2 - 3 | 142 | 160 | ||
| AK219009.1 | - | 1 - 2 | 84 | 160 | ||
| AK220121.1 | - | 1 - 2 | 84 | 160 | ||
| BC075617.1 | - | 1 - 2 | 112 | 5718202 | 160 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158750.1, AK170459.1, BC075617.1, NM_023431.5 transcripts.
Set Membership: |
|

Mum1
Genomic Map