Assay Details
| Gene Symbol: Mum1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023431.5 | NP_075920.4 | 3 - 4 | 314 | 138 | |
| GenBank mRNA | AK154610.1 | - | 3 - 4 | 233 | 138 | |
| AK158750.1 | - | 3 - 4 | 314 | 138 | ||
| AK170459.1 | - | 3 - 4 | 233 | 138 | ||
| AK205449.1 | - | 2 - 3 | 175 | 138 | ||
| AK220121.1 | - | 2 - 3 | 175 | 138 | ||
| BC075617.1 | - | 2 - 3 | 203 | 5718202 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mum1
Genomic Map