Assay Details
| Gene Symbol: Trappc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081055.1 | NP_001074524.1 | 2 - 3 | 291 | 83 | |
| GenBank mRNA | AK154290.1 | - | 2 - 3 | 227 | 83 | |
| AK166500.1 | - | 2 - 3 | 291 | 83 | ||
More Information
Set Membership: |
|

Trappc10
Genomic Map