Assay Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027425.3 | NP_081701.2 | 1 - 2 | 110 | 84 | |
| GenBank mRNA | AK045485.1 | - | 1 - 2 | 111 | 84 | |
| AK083205.1 | - | 1 - 2 | 95 | 84 | ||
| AK173195.1 | - | 1 - 2 | 71 | 84 | ||
| BB643897.1 | - | 1 - 2 | 111 | 84 | ||
| BC017645.1 | - | 1 - 2 | 72 | 84 | ||
| BC038265.1 | - | 1 - 2 | 51 | 84 | ||
| BC137736.1 | - | 1 - 2 | 59 | 8860755 | 84 | |
| BC137737.1 | - | 1 - 2 | 59 | 8860756 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083205.1, BC137736.1, BC137737.1, NM_027425.3 transcripts.
Set Membership: |
|

Rufy2
Genomic Map