Assay Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027425.3 | NP_081701.2 | 7 - 8 | 753 | 136 | |
| GenBank mRNA | AK011847.1 | - | 7 - 8 | 723 | 136 | |
| AK083205.1 | - | 7 - 8 | 738 | 136 | ||
| AK173195.1 | - | 7 - 8 | 714 | 136 | ||
| BC017645.1 | - | 7 - 8 | 715 | 136 | ||
| BC137736.1 | - | 7 - 8 | 702 | 8860755 | 136 | |
| BC137737.1 | - | 7 - 8 | 702 | 8860756 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rufy2
Genomic Map