| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000016283.mm.1 |
NM_001253857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016283.mm.1 |
NM_027384.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016283.mm.1 |
NM_027384.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016283.mm.1 |
TC1000002269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000016284.mm.1 |
NM_001253857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016284.mm.1 |
NM_027384.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016284.mm.1 |
NM_027384.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016284.mm.1 |
TC1000002269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000016286.mm.1 |
NM_001253857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016286.mm.1 |
NM_027384.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016286.mm.1 |
NM_027384.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016286.mm.1 |
TC1000002269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000016290.mm.1 |
NM_001253857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016290.mm.1 |
NM_027384.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016290.mm.1 |
NM_027384.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016290.mm.1 |
TC1000002269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000016285.mm.1 |
NM_001253857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016285.mm.1 |
NM_027384.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016285.mm.1 |
NM_027384.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016285.mm.1 |
TC1000002269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000016287.mm.1 |
NM_001253857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016287.mm.1 |
NM_027384.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016287.mm.1 |
NM_027384.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000016287.mm.1 |
TC1000002269.mm.1 |
MTA TC |