Assay Details
| Gene Symbol: Tet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253857.1 | NP_001240786.1 | 9 - 10 | 4851 | 63 | |
| NM_027384.1 | NP_081660.1 | 8 - 9 | 4755 | 63 | ||
| GenBank mRNA | AK129421.1 | - | 9 - 10 | 3104 | 63 | |
| GU079948.1 | - | 9 - 10 | 4851 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
inner cell mass cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
protein O-linked glycosylation
chromatin modification
stem cell population maintenance
regulation of DNA methylation
positive regulation of transcription from RNA polymerase II promoter
oxidation-reduction process
DNA demethylation
negative regulation of methylation-dependent chromatin silencing
regulation of genetic imprinting
DNA binding
iron ion binding
protein binding
zinc ion binding
oxidoreductase activity
structure-specific DNA binding
metal ion binding
dioxygenase activity
methylcytosine dioxygenase activity
inner cell mass cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
protein O-linked glycosylation
chromatin modification
stem cell population maintenance
regulation of DNA methylation
positive regulation of transcription from RNA polymerase II promoter
oxidation-reduction process
DNA demethylation
negative regulation of methylation-dependent chromatin silencing
regulation of genetic imprinting
DNA binding
iron ion binding
protein binding
zinc ion binding
oxidoreductase activity
structure-specific DNA binding
metal ion binding
dioxygenase activity
methylcytosine dioxygenase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123002 | GU079948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123002 | NM_027384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123002 | AK141438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122997 | NM_027384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | GU079948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | AK129421.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | NM_027384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | AI605989.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | AK010953.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | GU079948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | AK129421.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | NM_027384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | AI605989.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P512172 | AK010953.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920036 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299548 | AK122516.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299548 | BC056180.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299548 | NM_001081173.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523741 | AK050043.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523741 | AK132497.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523741 | AK148058.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523741 | AK163064.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523741 | AK167656.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523741 | AK168417.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523741 | NM_025884.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580636 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235759 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737076 | BC125621.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737076 | NM_146606.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737076 | X89689.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904965 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40947 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144799 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | GU079948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | AK129421.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | NM_027384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | AI605989.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | AK010953.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | GU079948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | AK129421.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | NM_027384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | AI605989.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P512172 | AK010953.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122997 | NM_027384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123002 | GU079948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123002 | NM_027384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123002 | AK141438.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07081445 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144799 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08789999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08287412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08208362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08823685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08834311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08834307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08721669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08562331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08542306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08603211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08562329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08531218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08789995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27188226 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10369695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369693 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369697 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369698 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419896_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419897_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455425_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429448_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369690 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369702 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369688 | - | - |
| Mouse Genome 430 2.0 Array | 1429448_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429448_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52463_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52463_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000016275.mm.1 | NM_001253857.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016275.mm.1 | NM_027384.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016275.mm.1 | NM_027384.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016275.mm.1 | TC1000002269.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000016278.mm.1 | NM_001253857.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016278.mm.1 | NM_027384.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016278.mm.1 | NM_027384.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000016278.mm.1 | TC1000002269.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429448_PM_s_at | - | - |
| Mouse Expression Set 430 | 1429448_s_at | - | - |
| Murine Genome U74v2 Set | 93186_at | - | - |
| Murine Genome U74v2 Set | 93186_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429448_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429448_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 552772 | - | - |
| ABI Mouse Genome Survey Microarray | 826419 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2598231 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2598231 | - | - |

Tet1
Genomic Map