Assay Details
| Gene Symbol: Dna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177372.3 | NP_796346.2 | 17 - 18 | 2771 | 71 | |
| GenBank mRNA | AK028381.1 | - | 17 - 18 | 2748 | 71 | |
| AK053856.1 | - | 2 - 3 | 1038 | 71 | ||
| AK087594.1 | - | 2 - 3 | 742 | 71 | ||
| AK129051.1 | - | 17 - 18 | 2756 | 71 | ||
| BC025182.1 | - | 12 - 13 | 1922 | 71 | ||
| BC115716.1 | - | 17 - 18 | 2725 | 40085888 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dna2
Genomic Map