Assay Details
| Gene Symbol: Dna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177372.3 | NP_796346.2 | 18 - 19 | 2858 | 125 | |
| GenBank mRNA | AK028381.1 | - | 18 - 19 | 2835 | 125 | |
| AK053856.1 | - | 3 - 4 | 1125 | 125 | ||
| AK087594.1 | - | 3 - 4 | 829 | 125 | ||
| AK129051.1 | - | 18 - 19 | 2843 | 125 | ||
| BC025182.1 | - | 13 - 14 | 2009 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dna2
Genomic Map