Assay Details
| Gene Symbol: Dna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177372.3 | NP_796346.2 | 2 - 3 | 321 | 66 | |
| GenBank mRNA | AK028381.1 | - | 2 - 3 | 298 | 66 | |
| AK129051.1 | - | 2 - 3 | 306 | 66 | ||
| BC030382.1 | - | 2 - 3 | 263 | 66 | ||
| BC115716.1 | - | 2 - 3 | 275 | 40085888 | 66 | |
| BY768981.1 | - | 2 - 3 | 323 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dna2
Genomic Map