| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000018226.mm.1 |
NM_001163276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018226.mm.1 |
NM_001163276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018226.mm.1 |
NM_198107.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018226.mm.1 |
NM_198107.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018226.mm.1 |
TC1000002484.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000018218.mm.1 |
NM_001163276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018218.mm.1 |
NM_001163276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018218.mm.1 |
NM_198107.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018218.mm.1 |
NM_198107.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018218.mm.1 |
TC1000002484.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000018220.mm.1 |
NM_001163276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018220.mm.1 |
NM_001163276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018220.mm.1 |
NM_198107.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018220.mm.1 |
NM_198107.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000018220.mm.1 |
TC1000002484.mm.1 |
MTA TC |