Assay Details
| Gene Symbol: Tbx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023814.4 | NP_076303.3 | 3 - 4 | 1026 | 101 | |
| GenBank mRNA | AF306666.1 | - | 3 - 4 | 961 | 101 | |
| AK012723.1 | - | 1 - 2 | 79 | 101 | ||
| AK032660.1 | - | 3 - 4 | 1028 | 101 | ||
| BC145691.1 | - | 3 - 4 | 923 | 40130994 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbx18
Genomic Map