Assay Details
| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172926.3 | NP_766514.2 | 6 - 7 | 692 | 151 | |
| GenBank mRNA | AK046839.1 | - | 6 - 7 | 634 | 151 | |
| AK049371.1 | - | 6 - 7 | 671 | 151 | ||
| BC080799.1 | - | 6 - 7 | 677 | 4948538 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx14
Genomic Map