Assay Details
| Gene Symbol: Sipa1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081028.1 | NP_001074497.1 | 13 - 14 | 4135 | 82 | |
| GenBank mRNA | AK154121.1 | - | 13 - 14 | 4131 | 82 | |
| AK163313.1 | - | 10 - 11 | 2517 | 82 | ||
| BC095969.1 | - | 3 - 4 | 668 | 82 | ||
More Information
Set Membership: |
|

Sipa1l3
Genomic Map