Assay Details
| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172920.4 | NP_766508.3 | 19 - 20 | 1869 | 87 | |
| GenBank mRNA | AK031291.1 | - | 1 - 2 | 949 | 87 | |
| AK143845.1 | - | 20 - 21 | 1874 | 87 | ||
| BC031382.1 | - | 12 - 13 | 1047 | 87 | ||
| BC116782.1 | - | 1 - 2 | 437 | 87 | ||
| BC116784.1 | - | 1 - 2 | 437 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpy19l1
Genomic Map