Assay Details
| Gene Symbol: Trp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127262.1 | NP_001120734.1 | 1 - 2 | 187 | 77 | |
| NM_001127263.1 | NP_001120735.1 | 1 - 2 | 187 | 77 | ||
| NM_001127264.1 | NP_001120736.1 | 1 - 2 | 187 | 77 | ||
| NM_001127265.1 | NP_001120737.1 | 1 - 2 | 187 | 77 | ||
| NM_011641.2 | NP_035771.1 | 1 - 2 | 187 | 77 | ||
| GenBank mRNA | AB010152.1 | - | 1 - 2 | 161 | 77 | |
| AK048623.1 | - | 1 - 2 | 187 | 77 | ||
| AK137253.1 | - | 1 - 2 | 188 | 77 | ||
| BC090649.1 | - | 1 - 2 | 169 | 30645427 | 77 | |
| BC092537.1 | - | 1 - 2 | 187 | 30504012 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB010152.1, AK048623.1, AK137253.1, BC090649.1, BC092537.1, NM_001127262.1, NM_001127263.1, NM_001127264.1, NM_001127265.1, NM_011641.2 transcripts.
Set Membership: |
|

Trp63
Genomic Map