Assay Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291910.1 | NP_001278839.1 | 6 - 7 | 2746 | 69 | |
| NM_009624.3 | NP_033754.2 | 5 - 6 | 2969 | 69 | ||
| GenBank mRNA | AK156940.1 | - | 6 - 7 | 2748 | 69 | |
| U30602.1 | - | 5 - 6 | 2276 | 69 | ||
| Z50190.1 | - | 4 - 5 | 2245 | 69 | ||
More Information
Set Membership: |
|

Adcy9
Genomic Map