Assay Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291910.1 | NP_001278839.1 | 7 - 8 | 2858 | 68 | |
| NM_009624.3 | NP_033754.2 | 6 - 7 | 3081 | 68 | ||
| GenBank mRNA | U30602.1 | - | 6 - 7 | 2388 | 68 | |
| Z50190.1 | - | 5 - 6 | 2357 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy9
Genomic Map