Assay Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291910.1 | NP_001278839.1 | 8 - 9 | 3066 | 82 | |
| NM_009624.3 | NP_033754.2 | 7 - 8 | 3289 | 82 | ||
| GenBank mRNA | AK156940.1 | - | 8 - 9 | 3068 | 82 | |
| U30602.1 | - | 7 - 8 | 2596 | 82 | ||
| Z50190.1 | - | 6 - 7 | 2565 | 82 | ||
More Information
Set Membership: |
|

Adcy9
Genomic Map