Assay Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291910.1 | NP_001278839.1 | 10 - 11 | 3372 | 79 | |
| NM_009624.3 | NP_033754.2 | 9 - 10 | 3595 | 79 | ||
| GenBank mRNA | AK156940.1 | - | 10 - 11 | 3374 | 79 | |
| U30602.1 | - | 9 - 10 | 2902 | 79 | ||
| Z50190.1 | - | 8 - 9 | 2871 | 79 | ||
More Information
Set Membership: |
|

Adcy9
Genomic Map