Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600000081.mm.1 |
NM_029090.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000081.mm.1 |
NM_029090.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000081.mm.1 |
TC1600002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000080.mm.1 |
NM_029090.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000080.mm.1 |
NM_029090.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000080.mm.1 |
TC1600002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600016796.mm.1 |
NM_029090.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016796.mm.1 |
NM_029090.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016796.mm.1 |
TC1600002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000085.mm.1 |
NM_029090.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000085.mm.1 |
NM_029090.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000085.mm.1 |
TC1600002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000082.mm.1 |
NM_029090.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000082.mm.1 |
NM_029090.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000082.mm.1 |
TC1600002166.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000088.mm.1 |
NM_029090.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000088.mm.1 |
NM_029090.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000088.mm.1 |
TC1600002166.mm.1 |
MTA TC |