Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 14 - 15 | 2107 | 76 | |
| GenBank mRNA | AK044836.1 | - | 14 - 15 | 2090 | 76 | |
| AK052832.1 | - | 14 - 15 | 2070 | 76 | ||
| AK075871.1 | - | 14 - 15 | 2072 | 76 | ||
| AK166448.1 | - | 9 - 10 | 1402 | 76 | ||
| AK170784.1 | - | 10 - 11 | 1739 | 76 | ||
| AK172902.1 | - | 14 - 15 | 2060 | 76 | ||
| BC026877.1 | - | 4 - 5 | 441 | 76 | ||
| BC046558.1 | - | 8 - 9 | 1111 | 76 | ||
| BC069859.1 | - | 5 - 6 | 660 | 76 | ||
| BC085258.1 | - | 5 - 6 | 478 | 76 | ||
| BC089026.1 | - | 14 - 15 | 2044 | 30615298 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spidr
Genomic Map