Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 17 - 18 | 2551 | 74 | |
| GenBank mRNA | AK044836.1 | - | 17 - 18 | 2534 | 74 | |
| AK052832.1 | - | 17 - 18 | 2514 | 74 | ||
| AK075871.1 | - | 17 - 18 | 2516 | 74 | ||
| AK146053.1 | - | 17 - 18 | 2550 | 74 | ||
| AK170784.1 | - | 13 - 14 | 2183 | 74 | ||
| BC026877.1 | - | 7 - 8 | 885 | 74 | ||
| BC046558.1 | - | 11 - 12 | 1555 | 74 | ||
| BC069859.1 | - | 8 - 9 | 1104 | 74 | ||
| BC085258.1 | - | 8 - 9 | 922 | 74 | ||
| BC089026.1 | - | 17 - 18 | 2488 | 30615298 | 74 | |
More Information
Set Membership: |
|

Spidr
Genomic Map