Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 18 - 19 | 2667 | 74 | |
| GenBank mRNA | AK044836.1 | - | 18 - 19 | 2650 | 74 | |
| AK052832.1 | - | 18 - 19 | 2630 | 74 | ||
| AK075871.1 | - | 18 - 19 | 2632 | 74 | ||
| AK146053.1 | - | 18 - 19 | 2666 | 74 | ||
| AK170784.1 | - | 14 - 15 | 2299 | 74 | ||
| BC026877.1 | - | 8 - 9 | 1001 | 74 | ||
| BC046558.1 | - | 12 - 13 | 1671 | 74 | ||
| BC069859.1 | - | 9 - 10 | 1220 | 74 | ||
| BC085258.1 | - | 9 - 10 | 1038 | 74 | ||
| BC089026.1 | - | 18 - 19 | 2604 | 30615298 | 74 | |
More Information
Set Membership: |
|

Spidr
Genomic Map