Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 2 - 3 | 258 | 96 | |
| GenBank mRNA | AK044836.1 | - | 2 - 3 | 241 | 96 | |
| AK052832.1 | - | 2 - 3 | 223 | 96 | ||
| AK075871.1 | - | 2 - 3 | 223 | 96 | ||
| AK146053.1 | - | 2 - 3 | 258 | 96 | ||
| BC089026.1 | - | 2 - 3 | 195 | 30615298 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052832.1 transcripts.
Set Membership: |
|

Spidr
Genomic Map