Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 7 - 8 | 1013 | 123 | |
| GenBank mRNA | AK044836.1 | - | 7 - 8 | 996 | 123 | |
| AK052832.1 | - | 7 - 8 | 976 | 123 | ||
| AK075871.1 | - | 7 - 8 | 978 | 123 | ||
| AK146053.1 | - | 7 - 8 | 1013 | 123 | ||
| AK170784.1 | - | 3 - 4 | 645 | 123 | ||
| AK172902.1 | - | 7 - 8 | 966 | 123 | ||
| BC089026.1 | - | 7 - 8 | 950 | 30615298 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spidr
Genomic Map