Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 9 - 10 | 1417 | 115 | |
| GenBank mRNA | AK044836.1 | - | 9 - 10 | 1400 | 115 | |
| AK052832.1 | - | 9 - 10 | 1380 | 115 | ||
| AK075871.1 | - | 9 - 10 | 1382 | 115 | ||
| AK146053.1 | - | 9 - 10 | 1417 | 115 | ||
| AK170784.1 | - | 5 - 6 | 1049 | 115 | ||
| AK172902.1 | - | 9 - 10 | 1370 | 115 | ||
| BC046558.1 | - | 3 - 4 | 421 | 115 | ||
| BC089026.1 | - | 9 - 10 | 1354 | 30615298 | 115 | |
More Information
Set Membership: |
|

Spidr
Genomic Map