Assay Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190974.1 | NP_001177903.1 | 10 - 11 | 1657 | 61 | |
| NM_001190975.1 | NP_001177904.1 | 9 - 10 | 1393 | 61 | ||
| NM_009465.4 | NP_033491.2 | 11 - 12 | 1684 | 61 | ||
| GenBank mRNA | AK033237.1 | - | 10 - 11 | 1626 | 61 | |
| AK155567.1 | - | 12 - 13 | 1570 | 61 | ||
| AK161294.1 | - | 4 - 5 | 357 | 61 | ||
| AK170010.1 | - | 10 - 11 | 1631 | 61 | ||
| AK170897.1 | - | 10 - 11 | 1290 | 61 | ||
| BC008551.1 | - | 6 - 7 | 774 | 61 | ||
| BC046618.1 | - | 11 - 12 | 1521 | 6494383 | 61 | |
| BC050914.1 | - | 11 - 12 | 1492 | 6418252 | 61 | |
| BC058230.1 | - | 11 - 12 | 1545 | 6313662 | 61 | |
| X63535.1 | - | 11 - 12 | 1433 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Axl
Genomic Map