Assay Details
| Gene Symbol: 4931429I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081121.1 | NP_001074590.1 | 5 - 6 | 1679 | 71 | |
| GenBank mRNA | AK016487.1 | - | 1 - 2 | 545 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

4931429I11Rik
Genomic Map