Assay Details
| Gene Symbol: 4931429I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081121.1 | NP_001074590.1 | 6 - 7 | 1968 | 81 | |
| GenBank mRNA | AK016487.1 | - | 2 - 3 | 834 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

4931429I11Rik
Genomic Map