Assay Details
| Gene Symbol: Gm5485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015373.1 | 2 - 3 | 71 | 91 | ||
| GenBank mRNA | AK078896.1 | - | 2 - 3 | 72 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | developmental pluripotency associated 2 | Dppa2 |
2410088E07Rik, C80932, D19Mgi18, ECAT15-2 |
-82.0kb | Anti-sense | Chr. 16 - 48310274 - 48319513 on NCBI Build GRCm38 |
| Downstream | microrchidia 1 | Morc1 | Morc | 19.0kb | Anti-sense | Chr. 16 - 48431237 - 48630905 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm5485
Genomic Map