| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900019085.mm.1 |
NM_001195681.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019085.mm.1 |
NM_001195681.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019085.mm.1 |
TC0900002287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019084.mm.1 |
NM_001195681.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019084.mm.1 |
NM_001195681.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019084.mm.1 |
TC0900002287.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019086.mm.1 |
NM_001195681.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019086.mm.1 |
NM_001195681.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019086.mm.1 |
TC0900002287.mm.1 |
MTA TC |