Assay Details
| Gene Symbol: Gm684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195681.1 | NP_001182610.1 | 3 - 4 | 365 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm684| Gene Symbol: Gm684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195681.1 | NP_001182610.1 | 3 - 4 | 365 | 64 | |
Set Membership: |
|