Assay Details
| Gene Symbol: Epha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010140.3 | NP_034270.1 | 1 - 2 | 196 | 77 | |
| GenBank mRNA | AK041935.1 | - | 1 - 2 | 197 | 77 | |
| AK045219.1 | - | 1 - 2 | 197 | 77 | ||
| AK084921.1 | - | 1 - 2 | 176 | 77 | ||
| AK089119.1 | - | 1 - 2 | 215 | 77 | ||
| AK132335.1 | - | 1 - 2 | 145 | 77 | ||
| BC060164.1 | - | 1 - 2 | 173 | 77 | ||
| BC093483.1 | - | 197 | 77 | |||
| BC138641.1 | - | 1 - 2 | 119 | 8861661 | 77 | |
| BC138643.1 | - | 1 - 2 | 119 | 8861663 | 77 | |
| BC145349.1 | - | 1 - 2 | 119 | 9053905 | 77 | |
| M68513.1 | - | 1 - 2 | 174 | 77 | ||
| M68515.1 | - | 1 - 2 | 174 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha3
Genomic Map