Assay Details
| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029436.3 | NP_083712.4 | 4 - 5 | 1349 | 80 | |
| GenBank mRNA | AK015239.1 | - | 4 - 5 | 1335 | 80 | |
| AK044892.1 | - | 4 - 5 | 1321 | 80 | ||
| AK046446.1 | - | 2 - 3 | 427 | 80 | ||
| AK163616.1 | - | 2 - 3 | 487 | 80 | ||
| BC021407.1 | - | 2 - 3 | 903 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl24
Genomic Map